1,2-Bis(dimethylamino)methan 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N,N,N',N'-Tetramethylmethylendiamin, 1,2-Di(dimethylamino)methan, N,N,N',N'-Tetramethyldiaminomethan
L04037.22
L04037.14
L04037.22EA
130
EUR
L04037.22
L04037.14
1,2-Bis(dimethylamino)methan 99%
1,2-Bis(dimethylamino)methan
Purity: 99%
CAS No.: 51-80-9
Molecular Formula: C5H14N2
Formula Weight: 102.18
Flash Point: -12°(10°F)
Refractive Index: 1.4010
Melting Point: -55°
Boiling Point: 83-85°
Beilstein Reference: 1731946
UN No.: 2733
MDL No.: MFCD00008328
Formel:
C₅H₁₄N₂ Molecular Weight: 102,18 g/mol Siedepunkt: 83…85 °C Schmelzpunkt: –55 °C Dichte: 0,749 Flammpunkt: –12 °C (10 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008328 CAS-Nummer: 51-80-9 EINECS: 200-124-7 UN: 2733 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...