1-Nonylisothiocyanat

Lieferant: FLUOROCHEM

009551-1G 009551-5G
FLUO009551-1GEA 55.4 EUR
FLUO009551-1G FLUO009551-5G
1-Nonylisothiocyanat
1-Nonylisothiocyanat
Formel: C₁₀H₁₉NS
Molecular Weight: 185.33 g/mol
Siedepunkt: 127…129 °C (10 mmHg)
Dichte: 0.89 g/cm³ (20 °C)
MDL: MFCD00041141
CAS-Nummer: 4430-43-7
UN: 2922
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR