1-Octylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: Octylsenföl

009554-5G 009554-25G
FLUO009554-5GEA 134 EUR
FLUO009554-5G FLUO009554-25G
1-Octylisothiocyanat
1-Octylisothiocyanat
Formel: C₉H₁₇NS
Molecular Weight: 171,31 g/mol
Siedepunkt: 123 °C (12 mmHg)
Dichte: 0,88 g/cm³ (20 °C)
MDL: MFCD00041139
CAS-Nummer: 4430-45-9
EINECS: 224-625-5
UN: 2922
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR