2-Methoxyethylisothiocyanat ≥98%

Lieferant: Thermo Scientific
Gefahr

L10839.06 L10839.03
L10839.06EA 94.3 EUR
L10839.06 L10839.03
2-Methoxyethylisothiocyanat ≥98%
2-Methoxyethylisothiocyanat

Purity: 98+% CAS No.: 38663-85-3 Molecular Formula: CH3O(CH2)2NCS Formula Weight: 117.17 Storage Temperature: 2°C to 8°C Flash Point: 66°(150°F) Refractive Index: 1.5120 Boiling Point: 88-92°/24mm Beilstein Reference: 773990 UN No.: 2922 MDL No.: MFCD00041126

Formel: C₄H₇NOS
Molecular Weight: 117,17 g/mol
Siedepunkt: 88…92 °C (24 mmHg)
Dichte: 1,080
Flammpunkt: 66 °C (150 °F)
MDL: MFCD00041126
CAS-Nummer: 38663-85-3
EINECS: 254-070-4
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR