2-Methoxyethylisothiocyanat

Lieferant: FLUOROCHEM

009506-1G 009506-5G 009506-25G
FLUO009506-1GEA 31.1 EUR
FLUO009506-1G FLUO009506-5G FLUO009506-25G
2-Methoxyethylisothiocyanat
2-Methoxyethylisothiocyanat
Formel: C₄H₇NOS
Molecular Weight: 117,17 g/mol
Siedepunkt: 88,5…92 °C (23.5 mmHg)
Dichte: 1,08 g/cm³ (20 °C)
MDL: MFCD00041126
CAS-Nummer: 38663-85-3
EINECS: 254-070-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR