1,6-Hexylendiisothiocyanat

Lieferant: FLUOROCHEM

008978-1G 008978-5G
FLUO008978-1GEA 218 EUR
FLUO008978-1G FLUO008978-5G
1,6-Hexylendiisothiocyanat
1,6-Hexylendiisothiocyanat
Formel: C₈H₁₂N₂S₂
MDL: MFCD00041137
CAS-Nummer: 5586-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR