Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) 99,8-100,1% (Trockenmasse) ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Aminomethylidintrimethanol, Tris(hydroxymethyl)methylamin, THAM, 2-Amino-2-(hydroxymethyl)propan-1,3-diol, Tromethamin, TRIS Base, 2-Amino-2-(hydroxymethyl)-1,3-propandiol
31801.18
31801.30
31801.A1
31801.18EA
27.3
EUR
31801.18
31801.30
31801.A1
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) 99,8-100,1% (Trockenmasse) ACS
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Purity: 99.8-100.1%
CAS No.: 77-86-1
Molecular Formula: (HOCH2)3CNH2
Formula Weight: 121.14
Physical Form: Crystalline
Density: 1.353
Melting Point: 171-172°
Boiling Point: 219-220°/10mm
Beilstein Reference: 741883
Merck Reference: 14,9772
MDL No.: MFCD00004679
Formel:
NH₂C(CH₂OH)₃ Molecular Weight: 121.14 g/mol Siedepunkt: 219…220 °C (10 mmHg) Schmelzpunkt: 171…172 °C Dichte: 1.353 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004679 CAS-Nummer: 77-86-1 EINECS: 201-064-4 Merck Index: 14,9772 |