Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) 99%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Aminomethylidintrimethanol, Tris(hydroxymethyl)methylamin, THAM, 2-Amino-2-(hydroxymethyl)propan-1,3-diol, Tromethamin, TRIS Base, 2-Amino-2-(hydroxymethyl)-1,3-propandiol
A18494.0E A18494.22 A18494.36
A18494.0EEA 290 EUR
A18494.0E A18494.22 A18494.36
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) 99%
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Purity: 99% CAS No.: 77-86-1 Molecular Formula: C4H11NO3 Formula Weight: 121.14 Melting Point: 168-172° Boiling Point: 219-220°/10mm Merck Reference: 14,9772 MDL No.: MFCD00004679
Formel: NH₂C(CH₂OH)₃
Molecular Weight: 121.14 g/mol
Siedepunkt: 219…220 °C (10 mmHg)
Schmelzpunkt: 168…172 °C
Dichte: 1.353
Lagertemperatur: Raumtemperatur
MDL: MFCD00004679
CAS-Nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM