Dibutylamin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-butyl-1-propanamin

A11671.0F A11671.AP
A11671.0FEA 101 EUR
A11671.0F A11671.AP
Dibutylamin ≥98%
Dibutylamin

Purity: 98+% CAS No.: 111-92-2 Molecular Formula: C8H19N Formula Weight: 129.25 Flash Point: 39°(102°F) Physical Form: Clear liquid Density: 0.767 Refractive Index: 1.4169 Appearance: Colourless Odour: Amine-like Melting Point: -62° Boiling Point: 159-160° Beilstein Reference: 506001 Merck Reference: 14,3032 UN No.: 2248 MDL No.: MFCD00009429

Formel: C₈H₁₉N
Molecular Weight: 129.25 g/mol
Siedepunkt: 159…160 °C
Schmelzpunkt: –62 °C
Dichte: 0.767
Flammpunkt: 39 °C (102 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009429
CAS-Nummer: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8 II
Merck Index: 13,03058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR