Dihexylamin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L06326.AE
L06326.AP
L06326.AEEA
48.4
EUR
L06326.AE
L06326.AP
Dihexylamin ≥98%
Dihexylamin
Purity: 98+%
CAS No.: 143-16-8
Molecular Formula: C12H27N
Formula Weight: 185.36
Flash Point: 104°(219°F)
Refractive Index: 1.4335
Melting Point: 3°
Boiling Point: 236°
Beilstein Reference: 1738519
UN No.: 2927
MDL No.: MFCD00009521
Formel:
C₁₂H₂₇N Molecular Weight: 185,35 g/mol Siedepunkt: 236 °C Schmelzpunkt: 3 °C Dichte: 0,790 Flammpunkt: 104 °C (219 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009521 CAS-Nummer: 143-16-8 EINECS: 205-588-4 UN: 2927 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...