Dipropylamin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
N-Propyl-1-propanamine
L15808.AE
L15808.AP
L15808.AEEA
24.6
EUR
L15808.AE
L15808.AP
Dipropylamin 99%
Dipropylamin
Purity: 99%
CAS No.: 142-84-7
Molecular Formula: C6H15N
Formula Weight: 101.19
Flash Point: 7°(45°F)
Refractive Index: 1.4050
Odour: Ammonia-like
Melting Point: -63°
Boiling Point: 109-110°
Beilstein Reference: 505974
Merck Reference: 14,3343
UN No.: 2383
MDL No.: MFCD00009362
Formel:
C₆H₁₅N Molecular Weight: 101.19 g/mol Siedepunkt: 109…110 °C Schmelzpunkt: –63 °C Dichte: 0.738 Flammpunkt: 7 °C (45 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009362 CAS-Nummer: 142-84-7 EINECS: 205-565-9 UN: 2383 ADR: 3 II Merck Index: 12,03406 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...