N-Methylethylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Aminoethylmethylamin, 1-Amino-2-(methylamino)ethan
M-Clarity™ Qualitätsstufe = MQ100
127019-100G
127019-25G
127019-5G
SIAL127019-100GEA
1020
EUR
SIAL127019-100G
SIAL127019-25G
SIAL127019-5G
N-Methylethylendiamin, Sigma-Aldrich®
N-Methylethylendiamin
Formel:
C₃H₁₀N₂ Molecular Weight: 74,13 g/mol Siedepunkt: 114 °C (1013 hPa) Dichte: 0,84 g/cm³ (20 °C) |
MDL:
MFCD00008165 CAS-Nummer: 109-81-9 EINECS: 203-707-4 UN: 2734 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...