2-(2-Aminoethylamino)ethanol 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-(2-Hydroxyethyl)ethylendiamin, AEEA

A10833.0E A10833.22 A10833.36
A10833.0EEA 125 EUR
A10833.0E A10833.22 A10833.36
2-(2-Aminoethylamino)ethanol 99%
2-(2-Aminoethylamino)ethanol

Purity: 99% CAS No.: 111-41-1 Molecular Formula: C4H12N2O Formula Weight: 104.15 Flash Point: 125°(257°F) Refractive Index: 1.4861 Odour: Amine-like pH: 11.8 Melting Point: -28° Boiling Point: 238-240° Beilstein Reference: 506012 UN No.: 2735 MDL No.: MFCD00008170

Formel: C₄H₁₂N₂O
Molecular Weight: 104,15 g/mol
Siedepunkt: 238…240 °C
Schmelzpunkt: –28 °C
Dichte: 1,029
Flammpunkt: 125 °C (257 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008170
CAS-Nummer: 111-41-1
EINECS: 203-867-5
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR