2-(Methylamino)ethanol 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Methyl-2-ethanolamin, N-Methyl-2-aminoethanol
B23180.AE
B23180.AP
B23180.AEEA
31.7
EUR
B23180.AE
B23180.AP
2-(Methylamino)ethanol 99%
2-(Methylamino)ethanol
Purity: 99%
CAS No.: 109-83-1
Molecular Formula: C5H7NO
Formula Weight: 75.11
Flash Point: 74°(165°F)
Refractive Index: 1.4400
Odour: Ammonia-like
Melting Point: -5°
Boiling Point: 159°
Beilstein Reference: 1071196
Merck Reference: 14,6015
UN No.: 2735
MDL No.: MFCD00002839
Formel:
C₃H₉NO Molecular Weight: 75,11 g/mol Siedepunkt: 159 °C Schmelzpunkt: –5 °C Dichte: 0,939 Flammpunkt: 74 °C (165 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002839 CAS-Nummer: 109-83-1 EINECS: 203-710-0 ADR: 8,III Merck Index: 14,6015 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...