BIS-TRIS-Propan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-(Propan-1,3-diyldiimino)bis[2-(hydroxymethyl)propan-1,3-diol], 1,3-Bis(tris(hydroxymethyl)methylamino)propan

M-Clarity™ Qualitätsstufe = MQ400

B6755-100G B6755-1KG B6755-250G B6755-25G B6755-500G
SIALB6755-100GEA 246 EUR
SIALB6755-100G SIALB6755-1KG SIALB6755-250G SIALB6755-25G SIALB6755-500G
BIS-TRIS-Propan, Sigma-Aldrich®
BIS-TRIS-Propan
Formel: C₁₁H₂₆N₂O₆
Molecular Weight: 282.34 g/mol
MDL: MFCD00004689
CAS-Nummer: 64431-96-5
EINECS: 264-899-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR