BIS-TRIS-Propan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-(Propan-1,3-diyldiimino)bis[2-(hydroxymethyl)propan-1,3-diol], 1,3-Bis(tris(hydroxymethyl)methylamino)propan

M-Clarity™ Qualitätsstufe = MQ400

B9410-100G B9410-250G B9410-25G
SIALB9410-100GEA 683 EUR
SIALB9410-100G SIALB9410-250G SIALB9410-25G
BIS-TRIS-Propan, Sigma-Aldrich®
BIS-TRIS-Propan
Formel: C₁₁H₂₆N₂O₆
Molecular Weight: 282.34 g/mol
MDL: MFCD00004689
CAS-Nummer: 64431-96-5
EINECS: 264-899-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR