2-(Cyclohexylamino)ethansulfonsäure (CHES)
Lieferant: Molekula
Synonyme:
N-Cyclohexyltaurine, CHES
15002366-500G
15002366-100G
MOLE15002366-500GEA
522
EUR
MOLE15002366-500G
MOLE15002366-100G
2-(Cyclohexylamino)ethansulfonsäure (CHES)
2-(Cyclohexylamino)ethansulfonsäure (CHES)
Formel:
C₈H₁₇NO₃S Molecular Weight: 207.29 g/mol Schmelzpunkt: 320 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003835 CAS-Nummer: 103-47-9 EINECS: 203-115-6 Merck Index: 13,02063 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...