2-(Cyclohexylamino)ethansulfonsäure (CHES), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Cyclohexyltaurine, CHES
M-Clarity™ Qualitätsstufe = MQ200
C2885-100G
C2885-1KG
C2885-250G
C2885-25G
SIALC2885-100GEA
284
EUR
SIALC2885-100G
SIALC2885-1KG
SIALC2885-250G
SIALC2885-25G
2-(Cyclohexylamino)ethansulfonsäure (CHES), Sigma-Aldrich®
2-(Cyclohexylamino)ethansulfonsäure (CHES)
Formel:
C₈H₁₇NO₃S Molecular Weight: 207.29 g/mol Schmelzpunkt: 320 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003835 CAS-Nummer: 103-47-9 EINECS: 203-115-6 Merck Index: 13,02063 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...