Ethylenglycolmonomethylether (2-Methoxyethanol) 99,30% ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methylglycol, Ethylenglycolmonomethylether
31733.AK
31733.K2
31733.AKEA
28.8
EUR
31733.AK
31733.K2
Ethylenglycolmonomethylether (2-Methoxyethanol) 99,30% ACS
Ethylenglycolmonomethylether (2-Methoxyethanol)
Purity: 99.3+%
CAS No.: 109-86-4
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 46°(115°F)
Physical Form: Liquid
Density: 0.965
Refractive Index: 1.4025
Appearance: Colourless
Odour: Characteristic
pH: 4-7
Melting Point: -85°
Boiling Point: 124-125°
Solubility: Miscible with water, alcohol, glycerol, ether, acetone, DMF
Beilstein Reference: 1731074
Merck Reference: 14,6038
UN No.: 1188
MDL No.: MFCD00002867
Formel:
C₃H₈O₂ Molecular Weight: 76,1 g/mol Siedepunkt: 124…125 °C Schmelzpunkt: –85 °C Dichte: 0,965 Flammpunkt: 46 °C (115 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002867 CAS-Nummer: 109-86-4 EINECS: 203-713-7 UN: 1188 ADR: 3,III Merck Index: 14,6038 |