Bis(2-ethoxyethyl)ether ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Diethylenglycoldiethylether, 2-Ethoxyethylether

A17478.0F A17478.AE A17478.AP
A17478.0FEA 145 EUR
A17478.0F A17478.AE A17478.AP
Bis(2-ethoxyethyl)ether ≥99%
Bis(2-ethoxyethyl)ether

Purity: 99% CAS No.: 112-36-7 Molecular Formula: C8H18O3 Formula Weight: 162.23 Flash Point: 71°(160°F) Refractive Index: 1.4120 Odour: Weak Melting Point: -44° Boiling Point: 188-190° Beilstein Reference: 1699259 Merck Reference: 14,3118 MDL No.: MFCD00009254

Formel: C₈H₁₈O₃
Molecular Weight: 162.23 g/mol
Siedepunkt: 188…190 °C
Schmelzpunkt: –44 °C
Dichte: 0.909
Flammpunkt: 71 °C (160 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009254
CAS-Nummer: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR