Aluminiumisopropoxid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Aluminiumtriisopropoxid, Aluminiumisopropylat, 2-Propanol Aluminiumsalz, Aluminiumisopropoxid, Aluminiumtriisopropylat
Purity: 98+%
CAS No.: 555-31-7
Molecular Formula: Al[OCH(CH3)2]3
Formula Weight: 204.25
Physical Form: Granular
Density: 1.035
Refractive Index: 1.0346
Sensitivity: Moisture Sensitive
Melting Point: 131-135°
Boiling Point: 135°/10mm
Solubility: Soluble in alcohol, benzene, and toluene. Decomposes in water
Merck Reference: 14,347
UN No.: 3181
MDL No.: MFCD00008870
Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates
Formel:
C₉H₂₁AlO₃ Molecular Weight: 204,25 g/mol Siedepunkt: 135 °C (10 mmHg) Schmelzpunkt: 131…135 °C Dichte: 1,035 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008870 CAS-Nummer: 555-31-7 EINECS: 209-090-8 UN: 3181 ADR: 4.1,II Merck Index: 14,347 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...