Natrium-1-decansulfonat
Lieferant: FLUOROCHEM
Synonyme:
Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt
009634-5G
009634-25G
FLUO009634-5GEA
34.1
EUR
FLUO009634-5G
FLUO009634-25G
Natrium-1-decansulfonat
Natrium-1-decansulfonat
Formel:
C₁₀H₂₁NaO₃S Molecular Weight: 244.33 g/mol Schmelzpunkt: > 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007526 CAS-Nummer: 13419-61-9 EINECS: 236-525-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...