Natrium-1-decansulfonat, Supelco®

Lieferant: Merck

Synonyme: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

M-Clarity™ Qualitätsstufe = MQ100

30631-10G-F 30631-50G-F
30631-10G-FEA 430 EUR
30631-10G-F 30631-50G-F
Natrium-1-decansulfonat, Supelco®
Natrium-1-decansulfonat
Formel: C₁₀H₂₁NaO₃S
Molecular Weight: 244,33 g/mol
Schmelzpunkt: > 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007526
CAS-Nummer: 13419-61-9
EINECS: 236-525-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b29000190d5d1de3ab217077367976219752cc0e&vid=9c9fdc7eafa642287007f4c1acb2e630628257a9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR