1-Hexansulfonsäure Natriumsalz, weiße Flocken
Lieferant: MP Biomedicals
Synonyme:
Natriumhexylsulfonat
Sodium hexanesulfonate is a low molecular weight alkylsulfonate that is used as an ion pairing reagent for HPLC and as an anionic surfactant. The anionic sulfonate counterion permits the separation and resolution of positively charged analytes. Because of its low molecular weight, it will not form micelles in solution and therefore has no CMC value.
For ion-pair chromatography. Sodium hexanesulfonate is utilized in the analysis of small organic compounds, and pharmaceutical products and metabolites by such techniques as HPLC.
- Presentation: White Flakes
- Soluble in water (100 mg/ml-clear, colorless), ethanol or methanol
- Storage temperature: Room Temperature
Formel:
C₆H₁₃O₃SNa Molecular Weight: 188,22 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007542 CAS-Nummer: 2832-45-3 EINECS: 220-601-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Reference |
Moisture | ≤10% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...