2-Propylvaleriansäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,2-Dipropylessigsäure, 2,2-Dipropylacetic acid, Valproinsäure, 2-Propylpentansäure
A12962.09
A12962.14
A12962.18
A12962.30
A12962.09EA
31.3
EUR
A12962.09
A12962.14
A12962.18
A12962.30
2-Propylvaleriansäure ≥98%
2-Propylvaleriansäure
Purity: 98+%
CAS No.: 99-66-1
Molecular Formula: C8H16O2
Formula Weight: 144.21
Flash Point: 126°(258°F)
Refractive Index: 1.4250
Odour: Weak, characteristic
Boiling Point: 219-220°
Beilstein Reference: 1750447
Merck Reference: 14,9913
UN No.: 3265
MDL No.: MFCD00002672
Formel:
C₈H₁₆O₂ Molecular Weight: 144.21 g/mol Siedepunkt: 219…220 °C Dichte: 0.905 Flammpunkt: 126 °C (258 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002672 CAS-Nummer: 99-66-1 EINECS: 202-777-3 UN: 3265 ADR: 8 III Merck Index: 12,10049 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...