5-Methylcapronsäure

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 5-Methylhexansäure

002773-5G 002773-25G
FLUO002773-5GEA 0 EUR
FLUO002773-5G FLUO002773-25G
5-Methylcapronsäure
5-Methylcapronsäure
Formel: C₇H₁₄O₂
Molecular Weight: 130,19 g/mol
MDL: MFCD00036494
CAS-Nummer: 628-46-6
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR