2,2-Dimethylbernsteinsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid

B24394.06 B24394.14
B24394.06EA 43.1 EUR
B24394.06 B24394.14
2,2-Dimethylbernsteinsäure ≥99%
2,2-Dimethylbernsteinsäure

Purity: 99% CAS No.: 597-43-3 Molecular Formula: C6H10O4 Formula Weight: 146.14 Melting Point: 139-142° Beilstein Reference: 1764476

Formel: C₆H₁₀O₄
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 139…142 °C
Dichte: 1,323 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004196
CAS-Nummer: 597-43-3
EINECS: 209-899-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR