1-Iod-5-(trifluormethyl)perfluorhexan
Lieferant: FLUOROCHEM
007076-25G
007076-5G
FLUO007076-25GEA
0
EUR
FLUO007076-25G
FLUO007076-5G
1-Iod-5-(trifluormethyl)perfluorhexan
1-Iod-5-(trifluormethyl)perfluorhexan
Formel:
C₇F₁₅I Molecular Weight: 495.96 g/mol |
MDL:
MFCD00042357 CAS-Nummer: 3486-08-6 EINECS: 222-475-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...