2-Methylbutylisothiocyanat

Lieferant: Thermo Scientific

TL00562DA TL00562.10 TL00562.50
MAYBTL00562DAEA 0 EUR
MAYBTL00562DA MAYBTL00562.10 MAYBTL00562.50
2-Methylbutylisothiocyanat
2-Methylbutylisothiocyanat
Formel: C₆H₁₁NS
Molecular Weight: 129.23 g/mol
MDL: MFCD00060414
CAS-Nummer: 4404-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR