2-Methylbutylisothiocyanat

Lieferant: FLUOROCHEM

009525-1G 009525-5G
FLUO009525-1GEA 60.9 EUR
FLUO009525-1G FLUO009525-5G
2-Methylbutylisothiocyanat
2-Methylbutylisothiocyanat
Formel: C₆H₁₁NS
Molecular Weight: 129.23 g/mol
MDL: MFCD00060414
CAS-Nummer: 4404-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR