2,2,4-Trimethyl-1-pentanol

Lieferant: CHEMPUR
Achtung

NRB02391.250 NRB02391.1
CHMPNRB02391.250EA 0 EUR
CHMPNRB02391.250 CHMPNRB02391.1
2,2,4-Trimethyl-1-pentanol
2,2,4-Trimethyl-1-pentanol
Formel: C₈H₁₈O
Molecular Weight: 130,23 g/mol
CAS-Nummer: 123-44-4
EINECS: 204-628-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR