2-Methyl-2-Butanol 99%, rein
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Amylenhydrat, tert-Pentylalkohol, tert-Amylalkohol, tert-Pentanol
166620010
166620025
166620100
ACRO166620010EA
101
EUR
ACRO166620010
ACRO166620025
ACRO166620100
2-Methyl-2-Butanol 99%, rein
2-Methyl-2-Butanol
Formel:
C₅H₁₂O Molecular Weight: 88,15 g/mol Siedepunkt: 102 °C (760 mmHg) Schmelzpunkt: –12 °C Dichte: 0,8 g/cm³ Flammpunkt: 20 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004478 CAS-Nummer: 75-85-4 EINECS: 200-908-9 UN: 1105 ADR: 3,II Merck Index: 13,07218 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4040 to 1.4060 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥98.5 % |
Sulfate (SO4) | ≤10 ppm |
Silicate (as SiO2) | ≤5 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Copper (Cu) | ≤0.1 ppm |
Iron (Fe) | ≤1 ppm |
Lead (Pb) | ≤0.05 ppm |
Magnesium (Mg) | ≤0.5 ppm |
Nickel (Ni) | ≤0.1 ppm |
Nitrogen (N) | ≤10 ppm |
Potassium (K) | ≤0.1 ppm |
Sodium (Na) | ≤0.5 ppm |
Zinc (Zn) | ≤0.1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...