3-Ethyl-3-pentanol ≥97%
Lieferant: Thermo Scientific
L03025.09
L03025.18
L03025.09EA
55.4
EUR
L03025.09
L03025.18
3-Ethyl-3-pentanol ≥97%
3-Ethyl-3-pentanol
Purity: 97%
CAS No.: 597-49-9
Molecular Formula: C7H16O
Formula Weight: 116.20
Flash Point: 40°(104°F)
Refractive Index: 1.4300
Odour: Characteristic
Melting Point: -13°
Boiling Point: 143°
Beilstein Reference: 1731643
UN No.: 1987
MDL No.: MFCD00004484
Formel:
C₇H₁₆O Molecular Weight: 116,2 g/mol |
MDL:
MFCD00004484 CAS-Nummer: 597-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...