Potassium-4-nitrophenyl sulphate ≥98% (durch TLC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Nitrophenyl sulphate, potassium salt, Potassium 4-Nitrophenyl sulphate, 4-Nitrophenyl sulphate potassium salt, sulphuric acid 4-Nitrophenyl ester potassium salt
M-Clarity™ Qualitätsstufe = MQ200
N3877-1G
N3877-500MG
N3877-5G
SIALN3877-1GEA
206
EUR
SIALN3877-1G
SIALN3877-500MG
SIALN3877-5G
Potassium-4-nitrophenyl sulphate ≥98% (durch TLC), Sigma-Aldrich®
Potassium-4-nitrophenyl sulphate
Chromogenic sulphatase substrate.
Solubility in water 50 mg/ml, clear.
Formel:
C₆H₄KNO₆S Molecular Weight: 257,257 g/mol Schmelzpunkt: 246…250 °C (Lit.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00007466 CAS-Nummer: 6217-68-1 EINECS: 228-288-5 |
Spezifikation Testergebnisse
Carbon | 27.4 - 28.6% |
Nitrogen | 27.4 - 28.6% |
Free p-Nitrophenol | ≤0.02% |
Potassium (K) | 14 - 16% |
Purity (TLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...