(α,α,α-Trifluor-3-tolyl)essigsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (3-(Trifluormethyl)phenyl)essigsäure

L10960.09 L10960.18
L10960.09EA 73.7 EUR
L10960.09 L10960.18
(α,α,α-Trifluor-3-tolyl)essigsäure 96%
(α,α,α-Trifluor-3-tolyl)essigsäure

Purity: 98+% CAS No.: 351-35-9 Molecular Formula: C9H7F3O2 Formula Weight: 204.15 Melting Point: 77-78° Beilstein Reference: 2213223

Formel: (C₆H₄(CF₃))CH₂COOH
Schmelzpunkt: 77…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004339
CAS-Nummer: 351-35-9
EINECS: 206-511-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR