(α,α,α-Trifluor-3-tolyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: (3-(Trifluormethyl)phenyl)essigsäure

002139-5G 002139-25G
FLUO002139-5GEA 32 EUR
FLUO002139-5G FLUO002139-25G
(α,α,α-Trifluor-3-tolyl)essigsäure
(α,α,α-Trifluor-3-tolyl)essigsäure
Formel: (C₆H₄(CF₃))CH₂COOH
Molecular Weight: 204,15 g/mol
MDL: MFCD00004339
CAS-Nummer: 351-35-9
EINECS: 206-511-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR