3-Cyclopentylpropionsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Cyclopentylpropanoic acid, 3-Cylcopentylpropanoic acid

111610250 111611000 111610050
ACRO111610250EA 46.6 EUR
ACRO111610250 ACRO111611000 ACRO111610050
3-Cyclopentylpropionsäure 98%
3-Cyclopentylpropionsäure
Formel: C₈H₁₄O₂
Molecular Weight: 142.2 g/mol
Siedepunkt: 130…132 °C (12 mmHg)
Dichte: 0.99 g/cm³
Flammpunkt: 46 °C
MDL: MFCD00001392
CAS-Nummer: 140-77-2
EINECS: 205-433-0
UN: 1993
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20°C, 589 nm)
Titration with NaOH ≥97.5 %
Absorbance ≤1.20 (1g/100ml cyclohexane) measured at the max. between 264nm and 276nm
Composition ≤0.5 % (delta 7-Stigmasterol)
Acid value ≤0.3 mg KOH/g
Peroxide value ≤10.0 meq O2/kg
Unsaponifiable matter ≤1.5 %
Sesame oil Complies

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR