Ethylindol-3-ylacetat ≥98%
Lieferant: Thermo Scientific
Synonyme:
3-Indolylessigsäureethylester, Indol-3-ylessigsäureethylester, Ethylindol-3-acetat, Indol-3-essigsäureethylester
A13474.06
A13474.14
AB148816.0100
A13474.06EA
62.1
EUR
A13474.06
A13474.14
ABCRAB148816.0100
Ethylindol-3-ylacetat ≥98%
Ethylindol-3-ylacetat
Purity: 98+%
CAS No.: 778-82-5
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Flash Point:
Formel:
C₁₂H₁₃NO₂ Molecular Weight: 203,24 g/mol Siedepunkt: 164…166 °C (1 mmHg) Schmelzpunkt: 41…44 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005635 CAS-Nummer: 778-82-5 EINECS: 212-296-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...