4-(3-Indolyl)butansäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Indol-3-buttersäure, 4-(Indol-3-yl)buttersäure
A15260.14
A15260.06
A15260.22
A15260.14EA
173
EUR
A15260.14
A15260.06
A15260.22
4-(3-Indolyl)butansäure ≥98%
4-(3-Indolyl)butansäure
Purity: 98%
CAS No.: 133-32-4
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Odour: Odourless
Melting Point: 121-125°
Beilstein Reference: 171120
Merck Reference: 14,4965
UN No.: 2811
MDL No.: MFCD00005664
Formel:
C₁₂H₁₃NO₂ Molecular Weight: 203.24 g/mol Schmelzpunkt: 121…125 °C Dichte: 1.27 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005664 CAS-Nummer: 133-32-4 EINECS: 205-101-5 UN: 2811 ADR: 6.1 III Merck Index: 13,04987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...