(3-Bromphenyl)essigsäure ≥98%

Lieferant: Thermo Scientific

Synonyme: 2-(3-Bromophenyl)acetic acid

B20758.06 B20758.14
B20758.06EA 110 EUR
B20758.06 B20758.14
(3-Bromphenyl)essigsäure ≥98%
(3-Bromphenyl)essigsäure

Purity: 98+% CAS No.: 1878-67-7 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 100-102° Beilstein Reference: 2045104

Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215,05 g/mol
Siedepunkt: 308 °C (1013 hPa)
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004330
CAS-Nummer: 1878-67-7
EINECS: 217-522-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR