(3-Bromphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(3-Bromophenyl)acetic acid

01134.25G 001134-5G 001134-25G
FLUO01134.25GEA 0 EUR
FLUO01134.25G FLUO001134-5G FLUO001134-25G
(3-Bromphenyl)essigsäure
(3-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215,05 g/mol
Siedepunkt: 308 °C (1013 hPa)
Schmelzpunkt: 102…103 °C
MDL: MFCD00004330
CAS-Nummer: 1878-67-7
EINECS: 217-522-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR