(4-Bromphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

001227-100G 001227-25G
FLUO001227-100GEA 97.2 EUR
FLUO001227-100G FLUO001227-25G
(4-Bromphenyl)essigsäure
(4-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215,05 g/mol
Siedepunkt: 326 °C (1013 hPa)
Schmelzpunkt: 94…98 °C
MDL: MFCD00004342
CAS-Nummer: 1878-68-8
EINECS: 217-523-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR