(2,4-Dichlorphenyl)essigsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DCAA
A11791.06
A11791.14
A11791.22
A11791.06EA
22.7
EUR
A11791.06
A11791.14
A11791.22
(2,4-Dichlorphenyl)essigsäure ≥98%
(2,4-Dichlorphenyl)essigsäure
Purity: 98+%
CAS No.: 19719-28-9
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Odour: Odourless
Melting Point: 128-130°
Beilstein Reference: 1948767
MDL No.: MFCD00004318
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 316 Schmelzpunkt: 128…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004318 CAS-Nummer: 19719-28-9 EINECS: 243-248-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...