(2,6-Dichlorphenyl)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Dichlorophenylacetic acid

B21354.06 B21354.14 B21354.22
B21354.06EA 23.8 EUR
B21354.06 B21354.14 B21354.22
(2,6-Dichlorphenyl)essigsäure ≥98%
(2,6-Dichlorphenyl)essigsäure

Purity: 98% CAS No.: 6575-24-2 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.04 Melting Point: 158-161° Beilstein Reference: 1952744

Formel: (C₆H₃Cl₂)CH₂COOH
Molecular Weight: 205,04 g/mol
Siedepunkt: 316 °C (1013 hPa)
Schmelzpunkt: 158…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004320
CAS-Nummer: 6575-24-2
EINECS: 229-504-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR