(2-Chlorphenyl)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chlorophenylacetic acid

A15834.09 A15834.18 A15834.30
A15834.09EA 34.3 EUR
A15834.09 A15834.18 A15834.30
(2-Chlorphenyl)essigsäure ≥98%
(2-Chlorphenyl)essigsäure

Purity: 98% CAS No.: 2444-36-2 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Odour: Odourless Melting Point: 94-97° Boiling Point: 145°/5mm Beilstein Reference: 1101523 MDL No.: MFCD00004317

Formel: (C₆H₄Cl)CH₂COOH
Molecular Weight: 170,6 g/mol
Siedepunkt: 145 °C (5 mmHg)
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004317
CAS-Nummer: 2444-36-2
EINECS: 219-482-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR