(3,4-Dichlorphenyl)essigsäure
Lieferant: FLUOROCHEM
Synonyme:
3,4-Dichlorophenylacetic acid
001384-25G
001384-100G
FLUO001384-25GEA
39.8
EUR
FLUO001384-25G
FLUO001384-100G
(3,4-Dichlorphenyl)essigsäure
(3,4-Dichlorphenyl)essigsäure
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 314 Schmelzpunkt: 91…92 °C |
MDL:
MFCD00004333 CAS-Nummer: 5807-30-7 EINECS: 227-368-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...