(3-Chlorphenyl)essigsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Chlorophenylacetic acid
A11210.06
A11210.14
A11210.22
A11210.06EA
28.8
EUR
A11210.06
A11210.14
A11210.22
(3-Chlorphenyl)essigsäure ≥98%
(3-Chlorphenyl)essigsäure
Purity: 98+%
CAS No.: 1878-65-5
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Odour: Odourless
Melting Point: 77-80°
Beilstein Reference: 2045103
MDL No.: MFCD00004332
Formel:
(C₆H₄Cl)CH₂COOH Molecular Weight: 170,6 g/mol Siedepunkt: 292 °C (1013 hPa) Schmelzpunkt: 77…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004332 CAS-Nummer: 1878-65-5 EINECS: 217-520-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...