(4-Chlorphenyl)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

A14974.0B A14974.18 A14974.30
A14974.0BEA 489 EUR
A14974.0B A14974.18 A14974.30
(4-Chlorphenyl)essigsäure ≥98%
(4-Chlorphenyl)essigsäure

Purity: 98% CAS No.: 1878-66-6 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Odour: Odourless Melting Point: 103-107° Beilstein Reference: 1072816 MDL No.: MFCD00004344

Formel: (C₆H₄Cl)CH₂COOH
Molecular Weight: 170,6 g/mol
Siedepunkt: 292 °C (1013 hPa)
Schmelzpunkt: 103…107 °C
Dichte: 1,43 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004344
CAS-Nummer: 1878-66-6
EINECS: 217-521-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR