(3-Chlor-2-fluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-(3-Chloro-2-fluorophenyl)acetic acid

009086-1G 009086-5G 009086-25G
FLUO009086-1GEA 34.1 EUR
FLUO009086-1G FLUO009086-5G FLUO009086-25G
(3-Chlor-2-fluorphenyl)essigsäure
(3-Chlor-2-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
MDL: MFCD01631554
CAS-Nummer: 261762-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR