(3-Chlor-4-fluorphenyl)essigsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Chloro-4-fluorophenylacetic acid
005666-1G
005666-5G
FLUO005666-1GEA
30.5
EUR
FLUO005666-1G
FLUO005666-5G
(3-Chlor-4-fluorphenyl)essigsäure
(3-Chlor-4-fluorphenyl)essigsäure
Formel:
(C₆H₃ClF)CH₂COOH Molecular Weight: 188,59 g/mol |
MDL:
MFCD01631555 CAS-Nummer: 705-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...